6-Hydroxyflavone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H0851-1G H0851-5G
TCH0851-001GEA 77.42 CAD
TCH0851-001G TCH0851-005G
6-Hydroxyflavone ≥98.0% (by HPLC, titration analysis)
6-Hydroxyflavone

CAS Number: 6665-83-4
MDL Number: MFCD00017329
Molecular Formula: C15H10O3
Molecular Weight: 238.24
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal

Formula: C₁₅H₁₀O₃
Boiling Pt: 395 °C (1013 hPa)
Melting Pt: 239 °C
MDL Number: MFCD00017329
CAS Number: 6665-83-4
EINECS: 229-704-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b554849ab567b3175a594003f452ef98933f936&vid=2b554849ab567b3175a594003f452ef98933f936&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR