tert-Butyl-N-hydroxycarbamate ≥98.0%
Supplier: TCI America
Synonyms:
N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine
H0848-25G
H0848-5G
TCH0848-25GEA
60.04
CAD
TCH0848-25G
TCH0848-005G
tert-Butyl-N-hydroxycarbamate ≥98.0%
tert-Butyl N-hydroxycarbamate
CAS Number: 36016-38-3
MDL Number: MFCD00002107
Molecular Formula: C5H11NO3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57
Other Product Names / Synonyms: N-Boc-hydroxylamine, N-(tert-Butoxycarbonyl)hydroxylamine, N-Hydroxycarbamic Acid tert-Butyl Ester
Formula:
C₅H₁₁NO₃ MW: 133.15 g/mol Melting Pt: 57 °C |
MDL Number:
MFCD00002107 CAS Number: 36016-38-3 EINECS: 252-836-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...