Hexylene di(acetate) ≥98.0%

Supplier: TCI America

H0832-25ML H0832-500ML
TCH0832-025MLEA 30.02 CAD
TCH0832-025ML TCH0832-500ML
Hexylene di(acetate) ≥98.0%
Hexylene di(acetate)

CAS Number: 6222-17-9
MDL Number: MFCD00191655
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Freezing point (°C): 9
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Hexamethylene Diacetate, 1,6-Hexanediol Diacetate

Formula: C₁₀H₁₈O₄
Boiling Pt: 260 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00191655
CAS Number: 6222-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1777e51cffbeb555ede734aa569e5fce67710591&vid=1777e51cffbeb555ede734aa569e5fce67710591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR