4-Quinazolone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Hydroxyquinazoline, Quinazolin-4(3H)-one

H0803-25G H0803-5G
TCH0803-025GEA 56.88 CAD
TCH0803-025G TCH0803-5G
4-Quinazolone ≥98.0% (by titrimetric analysis)
4-Quinazolone

CAS Number: 491-36-1
MDL Number: MFCD00511302
Molecular Formula: C8H6N2O
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 219


Other Product Names / Synonyms: 4-Quinazolinol

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 219 °C
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83ddbc15a1bd0667a9e3d3aa99f368c26ac34f27&vid=83ddbc15a1bd0667a9e3d3aa99f368c26ac34f27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR