5-Hydroxysophthalic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Hydroxyisophthalic acid
H0794-500G
H0794-25G
TCH0794-500GEA
984.34
CAD
TCH0794-500G
TCH0794-025G
5-Hydroxysophthalic acid ≥97.0% (by titrimetric analysis)
5-Hydroxysophthalic acid
CAS Number: 618-83-7
MDL Number: MFCD00002515
Molecular Formula: C8H6O5
Molecular Weight: 182.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Formula:
C₈H₆O₅ MW: 182.13 g/mol Boiling Pt: 386 °C (1013 hPa) Melting Pt: 298…301 °C |
MDL Number:
MFCD00002515 CAS Number: 618-83-7 EINECS: 210-565-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...