4'-Hydroxy-3'-methylacetophenone ≥98.0%

Supplier: TCI America

H0791-25G H0791-5G
TCH0791-025GEA 172.22 CAD
TCH0791-025G TCH0791-005G
4'-Hydroxy-3'-methylacetophenone ≥98.0%
4'-Hydroxy-3'-methylacetophenone

CAS Number: 876-02-8
MDL Number: MFCD00002231
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 108


Other Product Names / Synonyms: 4-Acetyl-2-methylphenol

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 180 °C
Melting Pt: 108 °C
MDL Number: MFCD00002231
CAS Number: 876-02-8
EINECS: 212-880-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37175da3a346c9d08741e6cdd863bc7252c64dd5&vid=37175da3a346c9d08741e6cdd863bc7252c64dd5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR