2'-Hydroxy-5'-methylacetophenone ≥98.0%

Supplier: TCI America

H0790-25G H0790-5G
TCH0790-025GEA 202.57 CAD
TCH0790-025G TCH0790-005G
2'-Hydroxy-5'-methylacetophenone ≥98.0%
2'-Hydroxy-5'-methylacetophenone

CAS Number: 1450-72-2
MDL Number: MFCD00002380
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 47
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Acetyl-4-methylphenol

Formula: C₉H₁₀O₂
Boiling Pt: 114 °C
Melting Pt: 47 °C
Density: 1.0797 g/cm³ (53 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002380
CAS Number: 1450-72-2
EINECS: 215-915-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a4a6e6c7e7faaf5cf0e7fc53fe4fc60c662caa&vid=62a4a6e6c7e7faaf5cf0e7fc53fe4fc60c662caa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR