2'-Hydroxy-4'-methoxyacetophenone ≥98.0%
Supplier: TCI America
H0789-5G
H0789-25G
TCH0789-005GEA
34.76
CAD
TCH0789-005G
TCH0789-025G
2'-Hydroxy-4'-methoxyacetophenone ≥98.0%
2'-Hydroxy-4'-methoxyacetophenone
CAS Number: 552-41-0
MDL Number: MFCD00008730
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 50
Flash Point (°C): 110
Formula:
C₉H₁₀O₃ Boiling Pt: 154 °C Melting Pt: 50 °C Density: 1.1604 g/cm³ (40 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008730 CAS Number: 552-41-0 EINECS: 209-012-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...