3-Hydroxypicolinic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Hydroxypyridine-2-carboxylic acid

H0787-5G H0787-25G
TCH0787-005GEA 149.53 CAD
TCH0787-005G TCH0787-25G
3-Hydroxypicolinic acid ≥98.0%
3-Hydroxypicolinic acid

CAS Number: 874-24-8
MDL Number: MFCD00006294
Molecular Formula: C6H5NO3
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 204


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 3-Hydroxypicolinic Acid

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 204 °C
MDL Number: MFCD00006294
CAS Number: 874-24-8
EINECS: 212-859-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5b67a1bf81c77d1344705fb88e19e85c40c2b2a&vid=a5b67a1bf81c77d1344705fb88e19e85c40c2b2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR