4-(Hydroxymethyl)benzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H0769-25G
H0769-10G
TCH0769-25GEA
396.58
CAD
TCH0769-25G
TCH0769-010G
4-(Hydroxymethyl)benzoic acid ≥98.0% (by titrimetric analysis)
4-(Hydroxymethyl)benzoic acid
CAS Number: 3006-96-0
MDL Number: MFCD00017598
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 183
Formula:
C₈H₈O₃ Melting Pt: 183 °C |
MDL Number:
MFCD00017598 CAS Number: 3006-96-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...