3-Hydroxy-2-nitro-6-picoline ≥98.0%

Supplier: TCI America

H0762-25G H0762-5G
TCH0762-25GEA 309.05 CAD
TCH0762-25G TCH0762-5G
3-Hydroxy-2-nitro-6-picoline ≥98.0%
3-Hydroxy-2-nitro-6-picoline

CAS Number: 15128-90-2
MDL Number: MFCD00006259
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 107


Other Product Names / Synonyms: 6-Methyl-2-nitro-3-pyridinol

Formula: C₆H₆N₂O₃
Melting Pt: 107 °C
MDL Number: MFCD00006259
CAS Number: 15128-90-2
EINECS: 239-192-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0915cb4becfeb956e93884e5b0fd2090c56dc489&vid=0915cb4becfeb956e93884e5b0fd2090c56dc489&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR