2,2,3,3,4,4-Hexafluoro-1,5-pentanediol ≥98.0%

Supplier: TCI America
Warning

H0746-1G H0746-5G H0746-25G
TCH0746-001GEA 104.28 CAD
TCH0746-001G TCH0746-005G 76220-114
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol ≥98.0%
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol

CAS Number: 376-90-9
MDL Number: MFCD00042434
Molecular Formula: C5H6F6O2
Molecular Weight: 212.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 79
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,5-Dihydroxy-2,2,3,3,4,4-hexafluoropentane

Formula: C₅H₆F₆O₂
MW: 212.09 g/mol
Boiling Pt: 112 °C
Melting Pt: 79 °C
Flash Pt: 110 °C
MDL Number: MFCD00042434
CAS Number: 376-90-9
EINECS: 206-819-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58d3d5de1bb98e8f5bb508cc2703168ed09e7459&vid=d5799419f93babdad2e73d87c588ba8113779658&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR