Hexafluoroglutaric anhydride ≥97.0%

Supplier: TCI America
Warning

Synonyms: 3,3,4,4,5,5-Hexafluorodihydro-2H-pyran-2,6(3H)-dione

H0745-5G H0745-25G
TCH0745-5GEA 113.76 CAD
TCH0745-5G TCH0745-001G TCH0745-25G
Hexafluoroglutaric anhydride ≥97.0%
Hexafluoroglutaric anhydride

CAS Number: 376-68-1
MDL Number: MFCD00006680
Molecular Formula: C5F6O3
Molecular Weight: 222.04
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 72
Specific Gravity (20/20): 1.65


Other Product Names / Synonyms: Hexafluoroglutaric Anhydride

Formula: C₅F₆O₃
MW: 222.04 g/mol
Boiling Pt: 80
Density: 1.65 (20 °C)
MDL Number: MFCD00006680
CAS Number: 376-68-1
EINECS: 206-811-8
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f18321cb0f8944d817ae86b487e3e8730cd44d9&vid=2f18321cb0f8944d817ae86b487e3e8730cd44d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR