5-Nitrovanillin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0728-25G H0728-10G H0728-5G
TCH0728-25GEA 107.44 CAD
TCH0728-25G TCH0728-010G TCH0728-5G
5-Nitrovanillin ≥98.0% (by titrimetric analysis)
5-Nitrovanillin

CAS Number: 6635-20-7
MDL Number: MFCD00007118
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 176


Other Product Names / Synonyms: 4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

Formula: C₈H₇NO₅
Melting Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00007118
CAS Number: 6635-20-7
EINECS: 229-633-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33948b9ed93287932d4fe07048a0e73b350b0291&vid=3ceb7982d93f19cbc438138479fc6e21c3896ee6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR