2-(4-Hydroxyphenyl)ethanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol

H0720-25G H0720-500G H0720-100G
TCH0720-025GEA 84.75 CAD
TCH0720-025G TCH0720-500G TCH0720-100G
2-(4-Hydroxyphenyl)ethanol ≥98.0%
2-(4-Hydroxyphenyl)ethanol

CAS Number: 501-94-0
MDL Number: MFCD00002902
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 91


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 4-(2-Hydroxyethyl)phenol, 4-Hydroxyphenethyl Alcohol, 2-(4-Hydroxyphenyl)ethyl Alcohol, Tyrosol

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 310
Melting Pt: 91 °C
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e61c90f272d5804f309f426e8423189f851c80b&vid=5e61c90f272d5804f309f426e8423189f851c80b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR