(R)-(-)-Methyl-3-hydroxybutyrate ≥98.0%

Supplier: TCI America

Synonyms: (R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate

H0705-25ML H0705-10ML
TCH0705-25MLEA 105.86 CAD
TCH0705-25ML TCH0705-010ML
(R)-(-)-Methyl-3-hydroxybutyrate ≥98.0%
(R)-(-)-Methyl 3-hydroxybutyrate

CAS Number: 3976-69-0
MDL Number: MFCD00063289
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 73
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: -22.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals, Bioscience, Lipids
Other Product Names / Synonyms: (R)-(-)-3-Hydroxybutyric Acid Methyl Ester

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 75 °C
Density: 1.06 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00063289
CAS Number: 3976-69-0
EINECS: 223-610-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13dc94dbf83df60056cfc77889e95d4aa128702d&vid=13dc94dbf83df60056cfc77889e95d4aa128702d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR