(S)-(+)-Methyl-3-hydroxybutyrate ≥98.0%
Supplier: TCI America
CAS Number: 53562-86-0
MDL Number: MFCD00064461
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 73
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: 22.5 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals, Bioscience, Lipids
Other Product Names / Synonyms: (S)-(+)-3-Hydroxybutyric Acid Methyl Ester
Formula:
C₅H₁₀O₃ Boiling Pt: 76 °C Density: 1.06 (20 °C) Flash Pt: 73 °C |
MDL Number:
MFCD00064461 CAS Number: 53562-86-0 EINECS: 258-628-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...