3-Hydroxy-3-methyl butyric acid

Supplier: TCI America

Synonyms: 3-Hydroxyisovaleric acid, 3-Hydroxy-3-methylbutanoic acid

H0701-5G H0701-25G
TCH0701-005GEA 140.62 CAD
TCH0701-005G TCH0701-25G
3-Hydroxy-3-methyl butyric acid
3-Hydroxy-3-methyl butyric acid

CAS Number: 625-08-1
MDL Number: MFCD00059081
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 3-Hydroxy-3-methylbutyric Acid

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Density: 1.10 (20 °C)
MDL Number: MFCD00059081
CAS Number: 625-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR