4'-Hydroxybiphenyl-4-carboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid
CAS Number: 58574-03-1
MDL Number: MFCD00059078
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (T,GC)
Form: Crystal
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4'-Hydroxybiphenyl-4-carboxylic Acid
Formula:
C₁₃H₁₀O₃ MW: 214.22 g/mol |
MDL Number:
MFCD00059078 CAS Number: 58574-03-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...