5-Hexyn-1-ol ≥95.0%

Supplier: TCI America
Warning

H0687-25ML H0687-5ML
TCH0687-025MLEA 412.38 CAD
TCH0687-025ML TCH0687-005ML
5-Hexyn-1-ol ≥95.0%
5-Hexyn-1-ol

CAS Number: 928-90-5
MDL Number: MFCD00002980
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 70
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C

Formula: C₆H₁₀O
Boiling Pt: 75 °C
Density: 0.91 (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002980
CAS Number: 928-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deb2559bf84ebebf3c0d74a23911b56828291fc0&vid=deb2559bf84ebebf3c0d74a23911b56828291fc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR