16-Hydroxypalmitic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0675-1G H0675-5G
TCH0675-001GEA 123.24 CAD
TCH0675-001G TCH0675-005G
16-Hydroxypalmitic acid ≥98.0% (by titrimetric analysis)
16-Hydroxypalmitic acid

CAS Number: 506-13-8
MDL Number: MFCD00002750
Molecular Formula: C16H32O3
Molecular Weight: 272.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 99


Other Product Names / Synonyms: 16-Hydroxypalmitic Acid, Juniperic Acid

Formula: C₁₆H₃₂O₃
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 99 °C
MDL Number: MFCD00002750
CAS Number: 506-13-8
EINECS: 208-028-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR