3-Hydroxy-4-nitrobenzoic acid ≥96.0%

Supplier: TCI America
Warning

H0635-250G H0635-25G
TCH0635-250GEA 1017.52 CAD
TCH0635-250G TCH0635-025G
3-Hydroxy-4-nitrobenzoic acid ≥96.0%
3-Hydroxy-4-nitrobenzoic acid

CAS Number: 619-14-7
MDL Number: MFCD00007110
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal

Formula: C₇H₅NO₅
Melting Pt: 234…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00007110
CAS Number: 619-14-7
EINECS: 210-580-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df8de238bee1480cc4531525a168d61c191d4df3&vid=f87ad60b9219204ef7da3c87d5b9ae18d7786b5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR