3-Hydroxy-p-anisidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxy-4-anisidine, 5-Amino-2-methoxyphenol, 3-Hydroxy-4-methoxyaniline, 4-Amino-2-hydroxyanisole

H0634-5G H0634-25G
TCH0634-005GEA 39.5 CAD
TCH0634-005G TCH0634-025G
3-Hydroxy-p-anisidine ≥98.0% (by GC, titration analysis)
3-Hydroxy-p-anisidine

CAS Number: 1687-53-2
MDL Number: MFCD00010222
Molecular Formula: C7H9NO2
Molecular Weight: 139.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 5-Aminoguaiacol, 5-Amino-2-methoxyphenol

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 130 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010222
CAS Number: 1687-53-2
EINECS: 216-873-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26b874d82b90825dc81d4cbfb4ce8be045cf167f&vid=26b874d82b90825dc81d4cbfb4ce8be045cf167f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR