2-Iodoperfluoropropane ≥95.0%
Supplier: TCI America
Synonyms:
1,1,1,2,3,3,3-Heptafluoro-2-iodopropane, Heptafluoroisopropyl iodide, Heptafluoro-2-iodopropane, 2-Iodoheptafluoropropane
CAS Number: 677-69-0
MDL Number: MFCD00042089
Molecular Formula: C3F7I
Molecular Weight: 295.93
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 40
Specific Gravity (20/20): 2.10
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Heptafluoro-2-iodopropane, Perfluoroisopropyl Iodide
Formula:
C₃F₇I MW: 295.93 g/mol Boiling Pt: 40 °C Melting Pt: –56 °C Density: 2.10 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042089 CAS Number: 677-69-0 EINECS: 211-643-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...