6-Methoxy-m-cresol ≥98.0%

Supplier: TCI America
Warning

H0624-25G H0624-5G
TCH0624-25GEA 584.6 CAD
TCH0624-25G TCH0624-005G
6-Methoxy-m-cresol ≥98.0%
6-Methoxy-m-cresol

CAS Number: 1195-09-1
MDL Number: MFCD00040901
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 117
Freezing point (°C): 34


Other Product Names / Synonyms: 2-Hydroxy-4-methylanisole, Isocreosole, 5-Methylguaiacol

Formula: C₈H₁₀O₂
Boiling Pt: 117 °C
Melting Pt: 38 °C
Density: 1.0973 g/cm³ (15 °C)
MDL Number: MFCD00040901
CAS Number: 1195-09-1
EINECS: 214-791-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b911ef3bb7df72a6b85f00f1b460de7ff5f62f8b&vid=b911ef3bb7df72a6b85f00f1b460de7ff5f62f8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR