N-Hydroxysuccinimide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0623-25G H0623-500G H0623-100G
TCH0623-025GEA 78.16 CAD
TCH0623-025G TCH0623-500G TCH0623-100G
N-Hydroxysuccinimide ≥98.0% (by titrimetric analysis)
N-Hydroxysuccinimide

CAS Number: 6066-82-6
MDL Number: MFCD00005516
Molecular Formula: C4H5NO3
Molecular Weight: 115.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 98


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Bioscience, Crosslinkers
Other Product Names / Synonyms: HOSu, NHS

Formula: C₄H₅NO₃
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005516
CAS Number: 6066-82-6
EINECS: 228-001-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e1b5c7b9a8d5bb7074d39186e2df832062f28e8&vid=1e1b5c7b9a8d5bb7074d39186e2df832062f28e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR