1-Hydroxycyclohexyl phenyl ketone ≥98.0%

Supplier: TCI America

H0617-500G H0617-25G H0617-100G
TCH0617-500GEA 422.49 CAD
TCH0617-500G TCH0617-025G 76220-112
1-Hydroxycyclohexyl phenyl ketone ≥98.0%
1-Hydroxycyclohexyl phenyl ketone

CAS Number: 947-19-3
MDL Number: MFCD00059561
Molecular Formula: C13H16O2
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 49
Flash Point (°C): 150


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 1-Benzoylcyclohexanol

Formula: C₁₃H₁₆O₂
Melting Pt: 49 °C
Flash Pt: 150 °C
MDL Number: MFCD00059561
CAS Number: 947-19-3
EINECS: 213-426-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0d9838914d5968ca030d3cef640c6735be798e7&vid=a0d9838914d5968ca030d3cef640c6735be798e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR