6-Hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-hydroxynaphthalene-2-carboxylic acid

H0612-25G H0612-500G H0612-5G
TCH0612-025GEA 52.93 CAD
TCH0612-025G TCH0612-500G TCH0612-005G
6-Hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
6-Hydroxy-2-naphthoic acid

CAS Number: 16712-64-4
MDL Number: MFCD00060070
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 248


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 6-Hydroxy-2-naphthalenecarboxylic Acid

Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Melting Pt: 248 °C
MDL Number: MFCD00060070
CAS Number: 16712-64-4
EINECS: 240-759-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08b81307b87bf6046d7165ee6079af7eb21ee91e&vid=08b81307b87bf6046d7165ee6079af7eb21ee91e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR