3-(4-Hydroxyphenyl)propionic acid ≥98.0%
Supplier: TCI America
Synonyms:
3-(4-Hydroxyphenyl)propanoic acid, p-hydroxyl phenylpropanol, 3-(p-Hydroxyphenyl)-Propionic Acid
H0599-250G
H0599-25G
TCH0599-250GEA
467.68
CAD
TCH0599-250G
TCH0599-025G
3-(4-Hydroxyphenyl)propionic acid ≥98.0%
3-(4-Hydroxyphenyl)propionic acid
CAS Number: 501-97-3
MDL Number: MFCD00002778
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 130
Other Product Names / Synonyms: 4-Hydroxyhydrocinnamic Acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 208…210 °C (14 mmHg) Melting Pt: 130 °C Density: 1.336 g/cm³ (20 °C) |
MDL Number:
MFCD00002778 CAS Number: 501-97-3 EINECS: 207-931-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...