1-Iodoperfluoropropane ≥95.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

H0596-25G H0596-5G
TCH0596-25GEA 778.94 CAD
TCH0596-25G TCH0596-005G
1-Iodoperfluoropropane ≥95.0% stabilized
1-Iodoperfluoropropane

(stabilized with Copper chip)
CAS Number: 754-34-7
MDL Number: MFCD00001061
Molecular Formula: C3F7I
Molecular Weight: 295.93
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 41
Melting point (°C): -95
Specific Gravity (20/20): 2.06


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Heptafluoro-1-iodopropane, Perfluoropropyl Iodide

Formula: C₃F₇I
Boiling Pt: 41 °C
Melting Pt: –95 °C
Density: 2.06 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001061
CAS Number: 754-34-7
EINECS: 212-045-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acbfc5cdc81ebccd76ad1e5fec1d939a63331cf1&vid=48b5cd064be3137afabc18d0a1b737e4abf7a790&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR