Potassium 2,5-dihydroxybenzenesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Hydroquinonesulfonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulfonate, 2,5-Dihydroxybenzenesulfonic acid potassium sal, 2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate

H0593-25G H0593-500G
TCH0593-025GEA 36.92 CAD
TCH0593-025G TCH0593-500G
Potassium 2,5-dihydroxybenzenesulfonate ≥98.0% (by titrimetric analysis)
Potassium 2,5-dihydroxybenzenesulphonate

CAS Number: 21799-87-1
MDL Number: MFCD00007475
Molecular Formula: C6H6O5S
Molecular Weight: 228.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2,5-Dihydroxybenzenesulfonic Acid Potassium Salt, Potassium 2,5-Dihydroxybenzenesulfonate, Hydroquinonesulfonic Acid Potassium Salt

Formula: C₆H₅KO₅S
MW: 228.27 g/mol
MDL Number: MFCD00007475
CAS Number: 21799-87-1
EINECS: 244-584-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86a297df1dff02921ed92e7eb18637cb1f4b7c9&vid=e86a297df1dff02921ed92e7eb18637cb1f4b7c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR