4-Heptylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0588-25ML H0588-5ML
TCH0588-025MLEA 368.14 CAD
TCH0588-025ML TCH0588-5ML
4-Heptylaniline ≥98.0% (by GC, titration analysis)
4-Heptylaniline

CAS Number: 37529-27-4
MDL Number: MFCD00007928
Molecular Formula: C13H21N
Molecular Weight: 191.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 286
Flash Point (°C): 110
Specific Gravity (20/20): 0.92

Formula: C₁₃H₂₁N
Boiling Pt: 286 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00007928
CAS Number: 37529-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cba27038a06007fd94ba29b10df01916a2bc97b&vid=5cba27038a06007fd94ba29b10df01916a2bc97b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR