2-(4-Hydroxyphenylazo)benzoic acid ≥98.0%
Supplier: TCI America
Synonyms:
4-Hydroxyazobenzene-2'-carboxylic acid, 2-(4-Hydroxyphenylazo)benzoic acid, 4'-Hydroxyazobenzene-2-carboxylic acid, 2-((4-Hydroxyphenyl)diazenyl)benzoic acid, HABA
CAS Number: 1634-82-8
MDL Number: MFCD00002428
Molecular Formula: C13H10N2O3
Molecular Weight: 242.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 201
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: HABA, 2-(4-Hydroxyphenylazo)benzoic Acid, 4-Hydroxyazobenzene-2'-carboxylic Acid
Formula:
C₁₃H₁₀N₂O₃ MW: 242.23 g/mol Melting Pt: 201 °C |
MDL Number:
MFCD00002428 CAS Number: 1634-82-8 EINECS: 216-655-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...