4-Hydroxybutanone ≥95.0%

Supplier: TCI America

Synonyms: 4-Hydroxy-2-butanone, 4-Hydroxybutan-2-one

H0584-25ML H0584-500ML H0584-100ML
TCH0584-025MLEA 61.07 CAD
TCH0584-025ML TCH0584-500ML TCH0584-100ML
4-Hydroxybutanone ≥95.0%
4-Hydroxybutanone

CAS Number: 590-90-9
MDL Number: MFCD00059005
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 89
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 2-Hydroxyethyl Methyl Ketone

Formula: C₄H₈O₂
Boiling Pt: 170 °C
Density: 1.02 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00059005
CAS Number: 590-90-9
EINECS: 209-693-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24618134bb75112a51ce0d0f8b3860df9009f063&vid=33a48598f5f0dffa030750cb1854e31c4cac18e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR