(1-Heptyl)triphenylphosphonium bromide ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Heptyltriphenylphosphonium bromide
H0545-500G
H0545-25G
TCH0545-500GEA
733.12
CAD
TCH0545-500G
TCH0545-025G
(1-Heptyl)triphenylphosphonium bromide ≥98.0%
(1-Heptyl)triphenylphosphonium bromide
CAS Number: 13423-48-8
MDL Number: MFCD00050249
Molecular Formula: C25H30BrP
Molecular Weight: 441.39
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 176
Formula:
H₃C(CH₂)₆P(C₆H₅)₃Br MW: 441.39 g/mol Melting Pt: 176 °C |
MDL Number:
MFCD00050249 CAS Number: 13423-48-8 EINECS: 236-539-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...