Polyethylene glycol 300

Supplier: TCI America

H0543-25G H0543-500G
TCH0543-25GEA 47.49 CAD
TCH0543-25G TCH0543-500G
Polyethylene glycol 300
Polyethylene glycol 300

CAS Number: 25322-68-3
MDL Number: MFCD01779599
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 196
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: PEG 300

Formula: H(OCH₂CH₂)nOH
Melting Pt: –15…–10 °C
Density: 1.13 (20 °C)
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR