(1-Hexyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Hexyltriphenylphosphonium bromide

H0540-25G H0540-500G H0540-100G
TCH0540-025GEA 86.9 CAD
TCH0540-025G TCH0540-500G TCH0540-100G
(1-Hexyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(1-Hexyl)triphenylphosphonium bromide

CAS Number: 4762-26-9
MDL Number: MFCD00011860
Molecular Formula: C24H28BrP
Molecular Weight: 427.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202

Formula: H₃C(CH₂)₅P(C₆H₅)₃Br
MW: 427.36 g/mol
Melting Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00011860
CAS Number: 4762-26-9
EINECS: 225-301-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afcc5ae41b09f7b3a38d31aeccb687bfeea34a22&vid=afcc5ae41b09f7b3a38d31aeccb687bfeea34a22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR