9-Heptadecanone ≥98.0%

Supplier: TCI America

H0536-25G H0536-5G
TCH0536-025GEA 1191.32 CAD
TCH0536-025G TCH0536-5G
9-Heptadecanone ≥98.0%
9-Heptadecanone

CAS Number: 540-08-9
MDL Number: MFCD00009573
Molecular Formula: C17H34O
Molecular Weight: 254.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 110


Other Product Names / Synonyms: Di-n-octyl Ketone, Pelargone

Formula: C₁₇H₃₄O
MW: 254.46 g/mol
Melting Pt: 54 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009573
CAS Number: 540-08-9
EINECS: 208-734-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e49b7fa5476e6c9138c4878f1945b993770b35d7&vid=0b3a34676f2c359de088396df39c2fc6e3427db0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR