3-Hydroxy-4-methoxycinnamic acid ≥98.0%

Supplier: TCI America

Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

H0524-1G H0524-5G
TCH0524-001GEA 89.85 CAD
TCH0524-001G TCH0524-005G
3-Hydroxy-4-methoxycinnamic acid ≥98.0%
3-Hydroxy-4-methoxycinnamic acid

CAS Number: 537-73-5
MDL Number: MFCD00004391
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 228


Product Categories: Bioscience, Phenylpropanoids Other Product Names / Synonyms: Hesperetic Acid, Isoferulic Acid

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Melting Pt: 228 °C
MDL Number: MFCD00004391
CAS Number: 537-73-5
EINECS: 208-676-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b6f58bdd7490d89c3ab430f4ace1d7a3bdb514&vid=11b6f58bdd7490d89c3ab430f4ace1d7a3bdb514&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR