Isopropyl-4-hydroxybenzoate ≥99.0%

Supplier: TCI America

Synonyms: Isopropylparaben, Isopropyl parahydroxybenzoate, Isopropyl p-Hydroxybenzoate, 4-Hydroxybenzoic acid isopropyl ester, Isopropyl Paraben, Isopropyl 4-Hydroxybenzoate

H0494-500G H0494-25G H0494-100G
TCH0494-500GEA 344.44 CAD
TCH0494-500G TCH0494-025G TCH0494-100G
Isopropyl-4-hydroxybenzoate ≥99.0%
Isopropyl 4-hydroxybenzoate

CAS Number: 4191-73-5
MDL Number: MFCD00016468
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >99.0% (T)
Form: Powder
Boiling point (°C): 160
Melting point (°C): 86


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Isopropyl Ester, Isopropylparaben

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 160 °C
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00016468
CAS Number: 4191-73-5
EINECS: 224-069-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3592d551548ab1d70014ccdc0ae8aef28a323a7&vid=a3592d551548ab1d70014ccdc0ae8aef28a323a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR