n-Hexane ≥96.0%
Supplier: TCI America
|
Danger
|
H0490-500ML
TCH0490-500MLEA
159.58
CAD
TCH0490-500ML
n-Hexane ≥96.0%
n-Hexane
[for HPLC Solvent]
CAS Number: 110-54-3
MDL Number: MFCD00009520
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Melting point (°C): -95
Flash Point (°C): -22
Specific Gravity (20/20): 0.66
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 68 °C Melting Pt: –95 °C Density: 0.66 (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...