2-Hydroxybenzophenone ≥95.0%

Supplier: TCI America

Synonyms: (2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone

H0470-25G H0470-5G
TCH0470-025GEA 260.7 CAD
TCH0470-025G TCH0470-005G
2-Hydroxybenzophenone ≥95.0%
2-Hydroxybenzophenone

CAS Number: 117-99-7
MDL Number: MFCD00002216
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 37
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Benzoylphenol

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 148 °C
Melting Pt: 37 °C
Density: 1.03 g/cm³ (172 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002216
CAS Number: 117-99-7
EINECS: 204-226-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea6c5dc86ff9c4882466538b283dc8182f909d56&vid=ea6c5dc86ff9c4882466538b283dc8182f909d56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR