(±)-1-Heptyn-3-ol ≥97.0%
Supplier: TCI America
H0455-1ML
H0455-5ML
TCH0455-1MLEA
47.4
CAD
TCH0455-1ML
TCH0455-005ML
(±)-1-Heptyn-3-ol ≥97.0%
(±)-1-Heptyn-3-ol
CAS Number: 7383-19-9
MDL Number: MFCD00041597
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 73
Flash Point (°C): 65
Specific Gravity (20/20): 0.87
Formula:
C₇H₁₂O Boiling Pt: 73 °C Density: 0.87 (20 °C) Flash Pt: 65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00041597 CAS Number: 7383-19-9 UN: 1987 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...