1-Hexanesulfonic acid sodium salt 98,0

Supplier: TCI America

Synonyms: Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate

H0447-25G H0447-500G H0447-100G
TCH0447-25GEA 158.63 CAD
TCH0447-25G TCH0447-500G TCH0447-100G
1-Hexanesulfonic acid sodium salt 98,0
1-Hexanesulphonic acid sodium salt

CAS Number: 2832-45-3
MDL Number: MFCD00007542
Molecular Formula: C6H14O3S
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1-Hexanesulfonic Acid Sodium Salt, Hexylsulfonic Acid Sodium Salt

Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88099d5ccc6bbf4411a1a3aa63207a0745517854&vid=88099d5ccc6bbf4411a1a3aa63207a0745517854&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR