4-Hexyloxyphenol ≥98.0%

Supplier: TCI America

H0439-5G H0439-25G
TCH0439-005GEA 170.64 CAD
TCH0439-005G TCH0439-025G
4-Hexyloxyphenol ≥98.0%
4-Hexyloxyphenol

CAS Number: 18979-55-0
MDL Number: MFCD00002337
Molecular Formula: C12H18O2
Molecular Weight: 194.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 46
Flash Point (°C): 110


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Hydroquinone Monohexyl Ether

Formula: C₁₂H₁₈O₂
Boiling Pt: 112 °C
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00002337
CAS Number: 18979-55-0
EINECS: 242-713-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01326aa654d3af6f2a5871a6098a16d9dadde4b3&vid=01326aa654d3af6f2a5871a6098a16d9dadde4b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR