1-Adamantanemethanol ≥99.0%

Supplier: TCI America

Synonyms: 1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol

H0420-5G H0420-1G H0420-25G
TCH0420-5GEA 90.06 CAD
TCH0420-5G TCH0420-001G TCH0420-25G
1-Adamantanemethanol ≥99.0%
1-Adamantanemethanol

CAS Number: 770-71-8
MDL Number: MFCD00074751
Molecular Formula: C11H18O
Molecular Weight: 166.26
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 118


Other Product Names / Synonyms: 1-(Hydroxymethyl)adamantane

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 118 °C
MDL Number: MFCD00074751
CAS Number: 770-71-8
EINECS: 212-225-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fe4202f36bdd2d3357ecb878212c5dc6793fc04&vid=9fe4202f36bdd2d3357ecb878212c5dc6793fc04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR