Piperidin-4-ol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Piperidinol, 4-Hydroxypiperidine

H0413-25G H0413-100G
TCH0413-025GEA 75.84 CAD
TCH0413-025G TCH0413-100G
Piperidin-4-ol ≥98.0%
Piperidin-4-ol

CAS Number: 5382-16-1
MDL Number: MFCD00005999
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 88
Flash Point (°C): 107


Other Product Names / Synonyms: 4-Piperidinol

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 222
Melting Pt: 88 °C
Flash Pt: 107 °C
MDL Number: MFCD00005999
CAS Number: 5382-16-1
EINECS: 226-373-1
UN: 3263
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c79926b76a67dbc605fa4bc3cbf7496f62d7b2a&vid=6c79926b76a67dbc605fa4bc3cbf7496f62d7b2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR