HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.0%

Supplier: TCI America

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

H0396-25G H0396-500G
TCH0396-025GEA 36.34 CAD
TCH0396-025G TCH0396-500G
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.0%
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

[Good's buffer component for biological research]
CAS Number: 7365-45-9
MDL Number: MFCD00006158
Molecular Formula: C8H18N2O4S
Molecular Weight: 238.30
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 210


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: HEPES

Formula: C₈H₁₈N₂O₄S
Melting Pt: 210 °C
Density: 1.448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5712d9b5cbff92ce8695d879fb0c0bfe0f0c011&vid=f5712d9b5cbff92ce8695d879fb0c0bfe0f0c011&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR