N-Hydroxyphthalimide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

H0395-25G H0395-500G H0395-100G
TCH0395-025GEA 60.18 CAD
TCH0395-025G TCH0395-500G TCH0395-100G
N-Hydroxyphthalimide ≥99.0% (by titrimetric analysis)
N-Hydroxyphthalimide

CAS Number: 524-38-9
MDL Number: MFCD00005891
Molecular Formula: C8H5NO3
Molecular Weight: 163.13
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 231


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Oxidation, Peptide Synthesis
Other Product Names / Synonyms: Phthaloxime, NHPI

Formula: C₈H₅NO₃
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 231 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00005891
CAS Number: 524-38-9
EINECS: 208-358-1
Merck Index: 13,04864
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=293404b099cf1ab1caa1dd75678bc95c81f6a78b&vid=293404b099cf1ab1caa1dd75678bc95c81f6a78b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR