3-Hydroxyflavone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H0379-1G H0379-10G
TCH0379-001GEA 64.78 CAD
TCH0379-001G TCH0379-010G
3-Hydroxyflavone ≥98.0% (by HPLC, titration analysis)
3-Hydroxyflavone

CAS Number: 577-85-5
MDL Number: MFCD00006832
Molecular Formula: C15H10O3
Molecular Weight: 238.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 170


Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: Flavonol

Formula: C₁₅H₁₀O₃
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 170 °C
Density: 1.393 g/cm³ (20 °C)
MDL Number: MFCD00006832
CAS Number: 577-85-5
EINECS: 209-416-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb7764dfa08d40334252b99485932590a605b37&vid=beb7764dfa08d40334252b99485932590a605b37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR